2-Nitro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
2-Nitro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12310.03
also known as L12310-03
Price (USD)/ Each
78.40
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
78.40
Add to cart
2-Nitro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Catalog numberL12310.03
Price (USD)/ Each
78.40
-
Add to cart
Chemical Identifiers
CAS402-14-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)95.0-106.0?C
Appearance (Color)Yellow to orange to brown or dark purple to purple
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only