Thermo Scientific Chemicals

Cyclopropyl phenyl sulfide, 98+%, Thermo Scientific Chemicals

Catalog number: L12321.03
1 g, Each
Thermo Scientific Chemicals

Cyclopropyl phenyl sulfide, 98+%, Thermo Scientific Chemicals

Catalog number: L12321.03
1 g, Each
Quantity
Catalog number: L12321.03
also known as L12321-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14633-54-6
IUPAC Name
(cyclopropylsulfanyl)benzene
Molecular Formula
C9H10S
InChI Key
YIBKCPJOFAUAKY-UHFFFAOYSA-N
SMILES
C1CC1SC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5800-1.5840 @ 20?C
Assay (GC)
≥98.0%
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text