4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals
4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12342.03
also known as L12342-03
Price (USD)/ Each
48.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
48.50
Add to cart
4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals
Catalog numberL12342.03
Price (USD)/ Each
48.50
-
Add to cart
Chemical Identifiers
CAS3694-57-3
IUPAC Name1-(isothiocyanatomethyl)-4-methoxybenzene
Molecular FormulaC9H9NOS
InChI KeyIMFQYAJJXFXVMM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CN=C=S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear yellow
Assay (GC)≥92.0%
Refractive Index1.5915-1.6000 @ 20?C
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.
RUO – Research Use Only