Thermo Scientific Chemicals

4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals

Catalog number: L12342.06
5 g, Each
Thermo Scientific Chemicals

4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals

Catalog number: L12342.06
5 g, Each
Quantity
Catalog number: L12342.06
also known as L12342-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3694-57-3
IUPAC Name
1-(isothiocyanatomethyl)-4-methoxybenzene
Molecular Formula
C9H9NOS
InChI Key
IMFQYAJJXFXVMM-UHFFFAOYSA-N
SMILES
COC1=CC=C(CN=C=S)C=C1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear yellow
Assay (GC)
≥92.0%
Refractive Index
1.5915-1.6000 @ 20?C

Description

4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text