4-Amino-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals
4-Amino-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12346.03
also known as L12346-03
Price (USD)/ Each
88.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
88.50
Add to cart
4-Amino-2-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific Chemicals
Catalog numberL12346.03
Price (USD)/ Each
88.50
-
Add to cart
Chemical Identifiers
CAS654-70-6
IUPAC Name4-amino-2-(trifluoromethyl)benzonitrile
Molecular FormulaC8H5F3N2
InChI KeyPMDYLCUKSLBUHO-UHFFFAOYSA-N
SMILESNC1=CC=C(C#N)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (GC)≥96.0%
Melting Point (clear melt)136.0-146.0?C
FormPowder or lumps or fused solid or granules
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only