Thermo Scientific Chemicals

4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L12356.AE
100 mL, Each
Thermo Scientific Chemicals

4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L12356.AE
100 mL, Each
Quantity
Catalog number: L12356.AE
also known as L12356-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1746-23-2
IUPAC Name
1-tert-butyl-4-ethenylbenzene
Molecular Formula
C12H16
InChI Key
QEDJMOONZLUIMC-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(C=C)C=C1
Assay from Supplier's CofA
≥92.5%
Form
Liquid
Stabilizer
45-55 ppm (4-tert-butylcatechol, TBC)
Appearance (Color)
Clear, colorless
Color (APHA)
≤20

Description

4-tert-Butylstyrene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylstyrene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in the dark. Protect from heat. Incompatible with heat, light, free radical initiators, UV radiation. Do not store together.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text