Latrunculin A
Get the sharpest, brightest images. Try new Invitrogen™ SlowFade™ Glass antifade RIMS mountant with refractive index of 1.52. Learn more »
Latrunculin A
Invitrogen™

Latrunculin A

Latrunculin A disrupts microfilament organization in cultured cells by binding to monomeric G-actin in a 1:1 complex at submicromolar concentrations.Read more
Promo ImagePromotion
Promo code:RIRP25
Save on Imaging Essentials to Create Your Next Masterpiece
14% off your purchase of $499 or more on Invitrogen cell stains and mounting products Learn More
Have Questions?
Catalog NumberQuantity
L12370Promo Image100 μg
Catalog number L12370
Price (USD)
636.65
Online Exclusive
656.00
Save 19.35 (3%)
Each
Add to cart
Quantity:
100 μg
Price (USD)
636.65
Online Exclusive
656.00
Save 19.35 (3%)
Each
Add to cart
Latrunculin A disrupts microfilament organization in cultured cells by binding to monomeric G-actin in a 1:1 complex at submicromolar concentrations. Its physiological effects include inhibition of fertilization, early embryological development and phagocytic internalization of immune complexes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Molecular FormulaC22H31NO5S
Quantity100 μg
Recommended StorageStore in freezer (-5°C to -30°C)
Shipping ConditionRoom Temperature
Sub Cellular LocalizationActin, Cytoskeleton
Physical FormSolid
Product TypeInhibitor
Unit SizeEach
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection