Thermo Scientific Chemicals

2-Chloropropene, 99%, Thermo Scientific Chemicals

Catalog number: L12374.06
5 g, Each
Thermo Scientific Chemicals

2-Chloropropene, 99%, Thermo Scientific Chemicals

Catalog number: L12374.06
5 g, Each
Quantity
Catalog number: L12374.06
also known as L12374-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-98-2
IUPAC Name
2-chloroprop-1-ene
Molecular Formula
C3H5Cl
InChI Key
PNLQPWWBHXMFCA-UHFFFAOYSA-N
SMILES
CC(Cl)=C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.3920-1.3960 @ 20?C
Form
Liquid

Description

2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text