Thermo Scientific Chemicals

N-Benzoylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: L12377.06
5 g, Each
Thermo Scientific Chemicals

N-Benzoylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: L12377.06
5 g, Each
Quantity
Catalog number: L12377.06
also known as L12377-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-23-3
IUPAC Name
benzoylthiourea
Molecular Formula
C8H8N2OS
InChI Key
DQMWMUMCNOJLSI-UHFFFAOYSA-N
SMILES
NC(=S)NC(=O)C1=CC=CC=C1
Assay (HPLC)
≥97.5%
Appearance (Color)
White
Melting Point (clear melt)
172-177?C
Form
Crystalline powder

Description

N-Benzoylthiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylthiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text