Thermo Scientific Chemicals

3,5-Di-tert-butylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: L12386.06
5 g, Each
Thermo Scientific Chemicals

3,5-Di-tert-butylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: L12386.06
5 g, Each
Quantity
Catalog number: L12386.06
also known as L12386-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16225-26-6
IUPAC Name
3,5-di-tert-butylbenzoic acid
Molecular Formula
C15H22O2
InChI Key
NCTSLPBQVXUAHR-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(=CC(=C1)C(O)=O)C(C)(C)C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid or needles
Assay (Silylated GC)
≥98.5%

Description

3,5-Di-tert-butylbenzoic acid is used in the synthesis of 5,5-di-tert-butylanilinium sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butylbenzoic acid is used in the synthesis of 5,5-di-tert-butylanilinium sulfate.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text