Thermo Scientific Chemicals

1-Naphthyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L12418.09
10 g, Each
Thermo Scientific Chemicals

1-Naphthyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L12418.09
10 g, Each
Quantity
Catalog number: L12418.09
also known as L12418-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
551-06-4
IUPAC Name
1-isothiocyanatonaphthalene
Molecular Formula
C11H7NS
InChI Key
JBDOSUUXMYMWQH-UHFFFAOYSA-N
SMILES
S=C=NC1=C2C=CC=CC2=CC=C1
Clarity
2.5% w/v solution in acetone is transparent to almost transparent by JIS K 8001 5.2
Melting Point (clear melt)
52-58°C
Identification (FTIR)
Conforms
Form
Crystalline powder or powder
Appearance (Color)
White to cream to yellow

Description

1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

So
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text