Thermo Scientific Chemicals

Homophthalonitrile, tech. 80%, Thermo Scientific Chemicals

Catalog number: L12419.14
25 g, Each
Thermo Scientific Chemicals

Homophthalonitrile, tech. 80%, Thermo Scientific Chemicals

Catalog number: L12419.14
25 g, Each
Quantity
Catalog number: L12419.14
also known as L12419-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3759-28-2
IUPAC Name
2-(cyanomethyl)benzonitrile
Molecular Formula
C9H6N2
InChI Key
GKHSEDFDYXZGCG-UHFFFAOYSA-N
SMILES
N#CCC1=CC=CC=C1C#N
Appearance (Color)
White to cream to brown
Assay (GC)
≥75.0%
Form
Crystals or powder or crystalline powder or lumps
Transition Temperature
73.0-82.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text