1x1 image pixel for data collection
Trifluoroacetamidine, tech. 85%
Trifluoroacetamidine, tech. 85%
Trifluoroacetamidine, tech. 85%
Trifluoroacetamidine, tech. 85%
Thermo Scientific Chemicals

Trifluoroacetamidine, tech. 85%

CAS: 354-37-0 | C2H3F3N2 | 112.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number L12422.14
also known as L12422-14
Price (USD)
325.65
Special offer
Online exclusive
Ends: 26-Sep-2025
402.00
Save 76.35 (19%)
Each
In stock
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
325.65
Special offer
Online exclusive
Ends: 26-Sep-2025
402.00
Save 76.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS354-37-0
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥ 83.0% based on 1-H & 19-F NMR (UK Sourced Material)
Proton NMRConsistent with structure (UK Sourced Material)
CommentMaterial Sourced in UK and US
FormLiquid
Appearance (Color)Clear, colourless to pale yellow or pink
View more
Ask our AI about this Product
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

Solubility
Fully miscible in water.

Notes
Store at -20°C. Incompatible with oxidizing agents. Moisture Sensitive.
RUO – Research Use Only

General References:

  1. R E London; S A Gabel. Determination of membrane potential and cell volume by 19F NMR using trifluoroacetate and trifluoroacetamide probes. Biochemistry (Washington).1989, 28, (6), 2378-2382.
  2. P Midoux; P Wahl; J C Auchet; M Monsigny. Fluorescence quenching of tryptophan by trifluoroacetamide. Biochimica et Biophysica Acta.1984, 801, (1), 16-25.
Have questions about this product? Ask our AI assisted search.
I purchase my Applied Biosystems 7900HT Fast Real-Time PCR System. Can I still order user-interchangeable standard 96-well, 384-well, and TaqManLow Density Array blocks?
How long can the Applied Biosystems 7900HT Fast Real-Time PCR System run unattended, and how many plates can be processed in that time frame?
When doing routine maintenance on the Applied Biosystems 7900HT Fast Real-Time PCR System, I have noticed that in the background plate there are some wells that are above 2500 fluorescent intensity units. What does this mean?
I have optimized my assay on the ABI PRISM 7900HT SDS. Will I need to re-optimize my assay to run on a standard block on the Applied Biosystems 7900HT Fast Real-Time PCR System?
Is the Applied Biosystems 7900HT Fast Real-Time PCR System available with a standard 96-well or 384-well block?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
10251712Certificate of AnalysisAug 09, 2024L12422.22, L12422.06, L12422.14
R19J036Certificate of AnalysisMay 31, 2023L12422.22, L12422.06, L12422.14
10245334Certificate of AnalysisJan 11, 2023L12422.22, L12422.06, L12422.14
Y15H051Certificate of AnalysisApr 12, 2022L12422.22, L12422.06, L12422.14
W01D013Certificate of AnalysisDec 08, 2021L12422.22, L12422.06, L12422.14
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link