2'-Fluoropropiophenone, 99%, Thermo Scientific Chemicals
2'-Fluoropropiophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Fluoropropiophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
Catalog number L12423.09
also known as L12423-09
Price (USD)/ Each
77.60
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
77.60
Add to cart
2'-Fluoropropiophenone, 99%, Thermo Scientific Chemicals
Catalog numberL12423.09
Price (USD)/ Each
77.60
-
Add to cart
Chemical Identifiers
CAS446-22-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5025-1.5065 @ 20?C
FormLiquid
2'-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, methanol. Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only
Andrew J. Beaumont; Christopher Kiely; A. Denise Rooney. Synthesis of novel chiral quaternary phosphonium fluorides: reagents for simple asymmetric nucleophilic fluorination reactions. Journal of Fluorine Chemistry.2001, 108, (1), 47-50.