Thermo Scientific Chemicals

2'-Fluoropropiophenone, 99%, Thermo Scientific Chemicals

Catalog number: L12423.18
50 g, Each
Thermo Scientific Chemicals

2'-Fluoropropiophenone, 99%, Thermo Scientific Chemicals

Catalog number: L12423.18
50 g, Each
Quantity
Catalog number: L12423.18
also known as L12423-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
446-22-0
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5025-1.5065 @ 20?C
Form
Liquid

Description

2'-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, methanol. Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text