Thermo Scientific Chemicals

(S)-(+)-2-Octanol, 99%, Thermo Scientific Chemicals

Catalog number: L12425.06
5 g, Each
Thermo Scientific Chemicals

(S)-(+)-2-Octanol, 99%, Thermo Scientific Chemicals

Catalog number: L12425.06
5 g, Each
Quantity
Catalog number: L12425.06
also known as L12425-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6169-06-8
IUPAC Name
octan-2-ol
Molecular Formula
C8H18O
InChI Key
SJWFXCIHNDVPSH-UHFFFAOYNA-N
SMILES
CCCCCCC(C)O
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid
Optical Rotation
10 ± 1? (c=1 in ethanol)
Refractive Index
1.4245-1.4275 @ 20?C

Description

(S)-(+)-2-Octanol is used as chiral compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Octanol is used as chiral compounds.

Solubility
Soluble in water. (1.12 g/L) at 25°C

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text