Thermo Scientific Chemicals

L-Homoserine, 99%, Thermo Scientific Chemicals

Catalog number: L12427.06
5 g, Each
Thermo Scientific Chemicals

L-Homoserine, 99%, Thermo Scientific Chemicals

Catalog number: L12427.06
5 g, Each
Quantity
Catalog number: L12427.06
also known as L12427-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
672-15-1
IUPAC Name
(2S)-2-amino-4-hydroxybutanoic acid
Molecular Formula
C4H9NO3
InChI Key
UKAUYVFTDYCKQA-VKHMYHEASA-N
SMILES
N[C@@H](CCO)C(O)=O
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
-8.6 ± 1° (c=2 in water)
Form
Crystals or powder or crystalline powder

Description

L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text