Phthalamic acid, 99%, Thermo Scientific Chemicals
Phthalamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalamic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L12432.14
also known as L12432-14
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
Phthalamic acid, 99%, Thermo Scientific Chemicals
Catalog numberL12432.14
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS88-97-1
IUPAC Name2-carbamoylbenzoic acid
Molecular FormulaC8H7NO3
InChI KeyCYMRPDYINXWJFU-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only