Thermo Scientific Chemicals

4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals

Catalog number: L12436.03
1 g, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals

Catalog number: L12436.03
1 g, Each
Quantity
Catalog number: L12436.03
also known as L12436-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
368-90-1
IUPAC Name
[4-(trifluoromethyl)phenyl]hydrazine
Molecular Formula
C7H7F3N2
InChI Key
DBNLGTYGKCMLLR-UHFFFAOYSA-N
SMILES
NNC1=CC=C(C=C1)C(F)(F)F
Form
Crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow to orange or pale brown
Assay (GC)
≥94.0%
Melting Point (clear melt)
58-65?C

Description

4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text