4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals
4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L12436.06
also known as L12436-06
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
Add to cart
4-(Trifluoromethyl)phenylhydrazine, 95%, Thermo Scientific Chemicals
Catalog numberL12436.06
Price (USD)/ Each
99.65
Online exclusive
110.00 
Save 10.35 (9%)
-
Add to cart
Chemical Identifiers
CAS368-90-1
IUPAC Name[4-(trifluoromethyl)phenyl]hydrazine
Molecular FormulaC7H7F3N2
InChI KeyDBNLGTYGKCMLLR-UHFFFAOYSA-N
SMILESNNC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to orange or pale brown
Assay (GC)≥94.0%
Melting Point (clear melt)58-65?C
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)phenylhydrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only