3-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals
3-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L12458.14
also known as L12458-14
Price (USD)/ Each
177.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
177.00
Add to cart
3-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberL12458.14
Price (USD)/ Each
177.00
-
Add to cart
Chemical Identifiers
CAS932-77-4
IUPAC Name1-bromo-3-(chloromethyl)benzene
Molecular FormulaC7H6BrCl
InChI KeyUDKGXKYEWBGQCG-UHFFFAOYSA-N
SMILESClCC1=CC(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.5805-1.5855 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only