Vinyltriethylsilane, 97%, Thermo Scientific Chemicals
Vinyltriethylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinyltriethylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L12461.06
also known as L12461-06
Price (USD)/ Each
103.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
103.00
Add to cart
Vinyltriethylsilane, 97%, Thermo Scientific Chemicals
Catalog numberL12461.06
Price (USD)/ Each
103.00
-
Add to cart
Chemical Identifiers
CAS1112-54-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay from Supplier's CofA≥96.0%
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only