Thermo Scientific Chemicals

Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: L12469.14
25 g, Each
Thermo Scientific Chemicals

Sodium hydrogen L-tartrate, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: L12469.14
25 g, Each
Quantity
Catalog number: L12469.14
also known as L12469-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
526-94-3
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Optical Rotation
24.0 ± 1.0? (C=3 in water)
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text