(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L12484.14
also known as L12484-14
Price (USD)/ Each
188.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
188.00
Add to cart
(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
Catalog numberL12484.14
Price (USD)/ Each
188.00
-
Add to cart
Chemical Identifiers
CAS4574-04-3
IUPAC Nametrimethyl(tetradecyl)azanium chloride
Molecular FormulaC17H38ClN
InChI KeyCEYYIKYYFSTQRU-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Water Content (Karl Fischer Titration)≤1.0%
Assay from Supplier's CofA≥97.5%
Heavy Metals≤ 5ppm
FormCrystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only