Thermo Scientific Chemicals

Diethylene glycol divinyl ether, 98%, stab. 0.1% potassium hydroxide, Thermo Scientific Chemicals

Catalog number: L12501.09
10 g, Each
Thermo Scientific Chemicals

Diethylene glycol divinyl ether, 98%, stab. 0.1% potassium hydroxide, Thermo Scientific Chemicals

Catalog number: L12501.09
10 g, Each
Quantity
Catalog number: L12501.09
also known as L12501-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
764-99-8
IUPAC Name
{2-[2-(ethenyloxy)ethoxy]ethoxy}ethene
Molecular Formula
C8H14O3
InChI Key
SAMJGBVVQUEMGC-UHFFFAOYSA-N
SMILES
C=COCCOCCOC=C
GC/MS Analysis
Conforms
Appearance (Color)
Colorless to pale yellow
Assay from Supplier's CofA
≥97.5% (GC)
Form
Liquid
Identification (FTIR)
Conforms

Description

Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text