Thermo Scientific Chemicals

1,4-Bis(3-aminopropyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: L12508.22
100 g, Each
Thermo Scientific Chemicals

1,4-Bis(3-aminopropyl)piperazine, 98%, Thermo Scientific Chemicals

Catalog number: L12508.22
100 g, Each
Quantity
Catalog number: L12508.22
also known as L12508-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7209-38-3
IUPAC Name
3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine
Molecular Formula
C10H24N4
InChI Key
XUSNPFGLKGCWGN-UHFFFAOYSA-N
SMILES
NCCCN1CCN(CCCN)CC1
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.5005-1.5045 @ 20°C

Description

1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text