2-Bromophenyl isocyanate, 97%, Thermo Scientific Chemicals
2-Bromophenyl isocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromophenyl isocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L12521.06
also known as L12521-06
Price (USD)/ Each
119.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
119.00
Add to cart
2-Bromophenyl isocyanate, 97%, Thermo Scientific Chemicals
Catalog numberL12521.06
Price (USD)/ Each
119.00
-
Add to cart
Chemical Identifiers
CAS1592-00-3
IUPAC Name1-bromo-2-isocyanatobenzene
Molecular FormulaC7H4BrNO
InChI KeyGOOVAYJIVMBWPP-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.5820-1.5870 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only