N-Phenylbis(trifluoromethanesulfonimide) acts as a mild triflating reagent as well as a transparent strong electron-withdrawing p-type dopant in carbon nanotubes. It is also employed as a reactant for the preparation of amphoteric alfa-boryl aldehydes. It plays an important role in the enantioselective synthesis of the core ring skeleton of leucosceptroids A-D and in steroselective sulfoxidation. Further, it is used in the enantioselective synthesis of beta-amino acids through the Mannich reaction. In addition to this, it is used in the preparation of sphingosine 1-phosphate-1 receptor agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenylbis(trifluoromethanesulfonimide) acts as a mild triflating reagent as well as a transparent strong electron-withdrawing p-type dopant in carbon nanotubes. It is also employed as a reactant for the preparation of amphoteric alfa-boryl aldehydes. It plays an important role in the enantioselective synthesis of the core ring skeleton of leucosceptroids A-D and in steroselective sulfoxidation. Further, it is used in the enantioselective synthesis of beta-amino acids through the Mannich reaction. In addition to this, it is used in the preparation of sphingosine 1-phosphate-1 receptor agonists.
Solubility
Soluble in methanol. Slightly soluble in chloroform and ethyl acetate.
Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Convenient reagent for preparation of triflates of phenols and amines: Tetrahedron Lett., 4607 (1973); Org. Lett., 4, 1231 (2002); or of enol triflates from carbonyl compounds: Tetrahedron Lett., 24, 979 (1983); J. Org. Chem., 51, 277 (1986). For discussion of the reactivity of triflates, see Trifluoromethanesulfonic anhydride, A11767.
Konoura, K.; Fujii, H.; Imaide, S.; Gouda, H.; Hirayama, S.; Hirono, S.; Nagase, H. Transformation of naltrexone into mesembrane and investigation of the binding properties of its intermediate derivatives to opioid receptors. Bioorg. Med. Chem. 2015, 23 (3), 439-448.
Chirkin, E.; Atkatlian, W.; Porée, F. H. Chapter One-The Securinega Alkaloids. Alkaloids Chem. Biol. 2015, 74, 1-120.