Ethyl 2-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals
Ethyl 2-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L12532.06
also known as L12532-06
Price (USD)/ Each
258.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
258.00
Add to cart
Ethyl 2-isothiocyanatopropionate, 97%, Thermo Scientific Chemicals
Catalog numberL12532.06
Price (USD)/ Each
258.00
-
Add to cart
Specifications
Chemical Name or MaterialEthyl 2-isothiocyanatopropionate
CAS39574-16-8
Health Hazard 1H302+H312-H314-H331-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only