Thermo Scientific Chemicals

Sodium dihydrogen citrate, 99%, Thermo Scientific Chemicals

Catalog number: L12557.36
500 g, Each
Thermo Scientific Chemicals

Sodium dihydrogen citrate, 99%, Thermo Scientific Chemicals

Catalog number: L12557.36
500 g, Each
Quantity
Catalog number: L12557.36
also known as L12557-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18996-35-5
IUPAC Name
sodium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Molecular Formula
C6H7NaO7
InChI Key
HWPKGOGLCKPRLZ-UHFFFAOYSA-M
SMILES
[Na+].OC(=O)CC(O)(CC(O)=O)C([O-])=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with bases, reducing agents and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text