Thermo Scientific Chemicals

3-Hydroxypropionitrile, 97%, Thermo Scientific Chemicals

Catalog number: L12570.14
25 g, Each
Thermo Scientific Chemicals

3-Hydroxypropionitrile, 97%, Thermo Scientific Chemicals

Catalog number: L12570.14
25 g, Each
Quantity
Catalog number: L12570.14
also known as L12570-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-78-4
IUPAC Name
3-hydroxypropanenitrile
Molecular Formula
C3H5NO
InChI Key
WSGYTJNNHPZFKR-UHFFFAOYSA-N
SMILES
OCCC#N
Assay (GC)
≥96.0% (non-U.S. sourced material)
Assay from Supplier's CofA
≥96.0% (U.S. sourced material)
Refractive Index
1.4240-1.4280 @ 20°C (non-U.S. sourced material)
Appearance (Color)
Clear colorless to yellow to brown or pale pink
Form
Liquid

Description

3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text