trans-2-Octene, 97%, Thermo Scientific Chemicals
trans-2-Octene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Octene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12606.06
also known as L12606-06
Price (USD)/ Each
84.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
84.40
Add to cart
trans-2-Octene, 97%, Thermo Scientific Chemicals
Catalog numberL12606.06
Price (USD)/ Each
84.40
-
Add to cart
Chemical Identifiers
CAS13389-42-9
IUPAC Name(2E)-oct-2-ene
Molecular FormulaC8H16
InChI KeyILPBINAXDRFYPL-HWKANZROSA-N
SMILESCCCCC\C=C\C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4100-1.4160 @ 20?C
Identification (FTIR)Conforms
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only