Thermo Scientific Chemicals

2,4,6-Trimethylphenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L12611.03
1 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethylphenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L12611.03
1 g, Each
Quantity
Catalog number: L12611.03
also known as L12611-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2958-62-5
IUPAC Name
2-isocyanato-1,3,5-trimethylbenzene
Molecular Formula
C10H11NO
InChI Key
HJHZRZFONUPQAA-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(N=C=O)C(C)=C1
Melting Point (clear melt)
42-48?C
Assay (GC)
≥97.5%
Form
Crystalline or fused solid or lumps or powder or crystalline powder
Appearance (Color)
White to pale cream to pale yellow

Description

2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines, bases and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text