2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines, bases and heat.
RUO – Research Use Only
Christoph Grundmann; Judith M.Dean. Nitrile oxides. V. Stable aromatic nitrile oxides1,2. J. Org. Chem. 1965, 30,(8), 2809-2812.
John F.W.Keana; Anthony P.Guzikowski; David D.Ward; Claude Morat; Faith L.Van Nice. Potent hydrophilic dienophiles. Synthesis and aqueous stability of several 4-aryl- and sulfonated 4-aryl-1,2,4-triazoline-3,5-diones and their immobilization on silica gel. J. Org. Chem. 1983, 48,(16), 2654-2660.