Thermo Scientific Chemicals

Methyl 4-bromobutyrate, 97%, Thermo Scientific Chemicals

Catalog number: L12636.09
10 g, Each
Thermo Scientific Chemicals

Methyl 4-bromobutyrate, 97%, Thermo Scientific Chemicals

Catalog number: L12636.09
10 g, Each
Quantity
Catalog number: L12636.09
also known as L12636-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4897-84-1
IUPAC Name
methyl 4-bromobutanoate
Molecular Formula
C5H9BrO2
InChI Key
QAWFLJGZSZIZHO-UHFFFAOYSA-N
SMILES
COC(=O)CCCBr
Assay (GC)
>96.0%
Appearance (Color)
Clear, colorless to pale yellow
Refractive Index
1.4590-1.4650 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text