Thermo Scientific Chemicals

Triethyl citrate, 99%, Thermo Scientific Chemicals

Catalog number: L12639.14
25 g, Each
Thermo Scientific Chemicals

Triethyl citrate, 99%, Thermo Scientific Chemicals

Catalog number: L12639.14
25 g, Each
Quantity
Catalog number: L12639.14
also known as L12639-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-93-0
IUPAC Name
1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C12H20O7
InChI Key
DOOTYTYQINUNNV-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC
Appearance (Color)
Clear, colourless
Assay (GC)
> 98.5%
Refractive Index
1.4405 - 1.4435 @20?C
Form
liquid

Description

Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text