Triethyl citrate, 99%, Thermo Scientific Chemicals
Triethyl citrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl citrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1000 g
Catalog number L12639.30
also known as L12639-30
Price (USD)/ Each
32.65
Online exclusive
36.60 
Save 3.95 (11%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
36.60 
Save 3.95 (11%)
Add to cart
Triethyl citrate, 99%, Thermo Scientific Chemicals
Catalog numberL12639.30
Price (USD)/ Each
32.65
Online exclusive
36.60 
Save 3.95 (11%)
-
Add to cart
Chemical Identifiers
CAS77-93-0
IUPAC Name1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular FormulaC12H20O7
InChI KeyDOOTYTYQINUNNV-UHFFFAOYSA-N
SMILESCCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
Assay (GC)> 98.5%
Refractive Index1.4405 - 1.4435 @20?C
Formliquid
Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Dekyndt, B.; Verin, J.; Neut, C.; Siepmann, F.; Siepmann, J. How to easily provide zero order release of freely soluble drugs from coated pellets. Int. J. Pharm. 2015, 478 (1), 31-38.
  2. Goyanes, A.; Hatton, G. B.; Basit, A. W. A dynamic in vitro model to evaluate the intestinal release behaviour of modified-release corticosteroid products. J. Drug Delivery Sci. Technol. 2015, 25, 36-42.