Thermo Scientific Chemicals

1,2-Dibenzoylethylene, predominantly trans, 96%, Thermo Scientific Chemicals

Catalog number: L12643.06
5 g, Each
Thermo Scientific Chemicals

1,2-Dibenzoylethylene, predominantly trans, 96%, Thermo Scientific Chemicals

Catalog number: L12643.06
5 g, Each
Quantity
Catalog number: L12643.06
also known as L12643-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
959-28-4
IUPAC Name
(2E)-1,4-diphenylbut-2-ene-1,4-dione
Molecular Formula
C16H12O2
InChI Key
WYCXGQSQHAXLPK-VAWYXSNFSA-N
SMILES
O=C(\C=C\C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Yellow to pale orange
Assay (GC)
≥95.0%
Form
Crystals or powder or crystalline powder or lumps

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text