Vinyl iodide, tech. 85%, Thermo Scientific Chemicals
Vinyl iodide, tech. 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinyl iodide, tech. 85%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12656.03
also known as L12656-03
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
Add to cart
Vinyl iodide, tech. 85%, Thermo Scientific Chemicals
Catalog numberL12656.03
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS593-66-8
IUPAC Nameiodoethene
Molecular FormulaC2H3I
InChI KeyGHXZPUGJZVBLGC-UHFFFAOYSA-N
SMILESIC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test10% w/v (in methanol): clear to slightly hazy
Identification (FTIR)Conforms
Solution Test10% w/v (in diethyl ether): clear to slightly hazy
Appearance (Color)Clear colorless to yellow to pink to red or red/orange
Assay (GC)≥83.0%
View more
Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to light. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Edward Piers.; Pierre C. Marais. A new five-membered ring annulation method based on palladium(0)-catalyzed intramolecular coupling of vinyl iodide and enolate anion functions. J. Org. Chem. 1990, 55 (11), 3454-3455.
  2. Z. M. Liu.; X. L. Zhou.; D. A. Buchanan.; J. Kiss, J. M. White. The surface chemistry of vinyl iodide on platinum(111). J. Am. Chem. Soc. 1992, 114 (6), 2031-2039.