Thermo Scientific Chemicals

9,10-Dibromoanthracene, 98%, Thermo Scientific Chemicals

Catalog number: L12680.14
25 g, Each
Thermo Scientific Chemicals

9,10-Dibromoanthracene, 98%, Thermo Scientific Chemicals

Catalog number: L12680.14
25 g, Each
Quantity
Catalog number: L12680.14
also known as L12680-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
523-27-3
IUPAC Name
9,10-dibromoanthracene
Molecular Formula
C14H8Br2
InChI Key
BRUOAURMAFDGLP-UHFFFAOYSA-N
SMILES
BrC1=C2C=CC=CC2=C(Br)C2=CC=CC=C12
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text