9,10-Dibromoanthracene, 98%, Thermo Scientific Chemicals
9,10-Dibromoanthracene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9,10-Dibromoanthracene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number L12680.22
also known as L12680-22
Price (USD)/ Each
303.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
303.00
Add to cart
9,10-Dibromoanthracene, 98%, Thermo Scientific Chemicals
Catalog numberL12680.22
Price (USD)/ Each
303.00
-
Add to cart
Chemical Identifiers
CAS523-27-3
IUPAC Name9,10-dibromoanthracene
Molecular FormulaC14H8Br2
InChI KeyBRUOAURMAFDGLP-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=C(Br)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dibromoanthracene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place
RUO – Research Use Only

General References:

  1. Jiro Tanaka.; Masao Shibata.Electronic Absorption and Emission Spectra of 9, 10-Dichloroanthracene and 9, 10-Dibromoanthracene Crystals. Bulletin of the Chemical Society of Japan. 1968, 48 (1), 34-41.
  2. W. Grant McGimpsey.; J. C. Scaiano. Photochemistry and photophysics from upper triplet levels of 9,10-dibromoanthracene. J. Am. Chem. Soc. 1989, 111 (1), 335-340.