Thermo Scientific Chemicals

D-Amygdalin hydrate, 96%, Thermo Scientific Chemicals

Catalog number: L12731.04
2 g, Each
Thermo Scientific Chemicals

D-Amygdalin hydrate, 96%, Thermo Scientific Chemicals

Catalog number: L12731.04
2 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L12731.04
also known as L12731-04
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
29883-15-6
IUPAC Name
2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile
Molecular Formula
C20H27NO11
InChI Key
XUCIJNAGGSZNQT-ARGBPYSKNA-N
SMILES
OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Appearance (Color)
White
Assay (HPLC)
≥95.0%
Loss on Drying
1.93-5.58% (110?C/constant weight) (0.5-1.5 waters)
Form
Crystals or powder or crystalline powder or fused solid
Optical Rotation
-37.0 ± 1.0? (C=2 in water, dry weight basis)

Description

A benzylic glycoside that has been used as an antineoplastic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell