Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L12739.AE
also known as L12739-AE
Price (USD)/ Each
41.90
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
41.90
Add to cart
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Catalog numberL12739.AE
Price (USD)/ Each
41.90
-
Add to cart
Chemical Identifiers
CAS106-63-8
IUPAC Name2-methylpropyl prop-2-enoate
Molecular FormulaC7H12O2
InChI KeyCFVWNXQPGQOHRJ-UHFFFAOYSA-N
SMILESCC(C)COC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Assay (GC)> 98.5%
Solution Test(10% v/v in methanol) : Clear
Refractive Index1.4135 - 1.4165 @ 20?C
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only