Thermo Scientific Chemicals

Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L12739.AP
500 mL, Each
Thermo Scientific Chemicals

Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L12739.AP
500 mL, Each
Quantity
Catalog number: L12739.AP
also known as L12739-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-63-8
IUPAC Name
2-methylpropyl prop-2-enoate
Molecular Formula
C7H12O2
InChI Key
CFVWNXQPGQOHRJ-UHFFFAOYSA-N
SMILES
CC(C)COC(=O)C=C
Appearance (Color)
Clear, colorless
Assay (GC)
> 98.5%
Solution Test
(10% v/v in methanol) : Clear
Refractive Index
1.4135 - 1.4165 @ 20?C
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text