Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
RUO – Research Use Only
S.S.Lord. Infrared analysis of toluene-2,4-diisocyanate and toluene-2,6-diisocyanate mixtures. Anal. Chem. 1957, 29,(4), 497-499.
C.Sango. Improved method for determination of traces of isocyanates in working atmospheres by high performance liquid chromatography. Journal of Liquid Chromatography. 1979, 2,(6), 763-774.