Thermo Scientific Chemicals

N,N-Di-n-butylformamide, 99%, Thermo Scientific Chemicals

Catalog number: L12755.36
500 g, Each
Thermo Scientific Chemicals

N,N-Di-n-butylformamide, 99%, Thermo Scientific Chemicals

Catalog number: L12755.36
500 g, Each
Quantity
Catalog number: L12755.36
also known as L12755-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
761-65-9
IUPAC Name
N,N-dibutylformamide
Molecular Formula
C9H19NO
InChI Key
NZMAJUHVSZBJHL-UHFFFAOYSA-N
SMILES
CCCCN(CCCC)C=O
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4425-1.4460 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text