Thermo Scientific Chemicals

2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%, Thermo Scientific Chemicals

Catalog number: L12760.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%, Thermo Scientific Chemicals

Catalog number: L12760.06
5 g, Each
Quantity
Catalog number: L12760.06
also known as L12760-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42712-64-1
IUPAC Name
2-amino-1,4-dihydroquinolin-4-one
Molecular Formula
C9H8N2O
InChI Key
LWGUCIXHBVVATR-UHFFFAOYSA-N
SMILES
NC1=CC(=O)C2=CC=CC=C2N1
Water Content (Karl Fischer Titration)
≤12%
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0% (dry wt. basis)
Appearance (Color)
White to pale brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text