4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only
Noritada Matsuo.; Andrew S. Kende. 1,1,2-Trifluoro-1,3-butadiene: a convenient C4 intermediate for functionalized monofluoroolefins. J. Org. Chem. 1988, 53 (10), 2 304-2308.
Dorothea Golze.; Maik Icker.; Stefan Berger. Implementation of two-qubit and three-qubit quantum computers using liquid-state nuclear magnetic resonance. Concepts in Magnetic Resonance Part A. 2012, 40A (1), 25-37.
Precursor, by dehydrobromination under phase-transfer conditions, of 1,1,2-trifluoro-1,3-butadiene, a convenient C4 intermediate for functionalized monofluoroolefins: J. Org. Chem., 53, 2304 (1988).