3-(4-Morpholinyl)propyl isothiocyanate, 96%, Thermo Scientific Chemicals
3-(4-Morpholinyl)propyl isothiocyanate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(4-Morpholinyl)propyl isothiocyanate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L12819.06
also known as L12819-06
Price (USD)/ Each
97.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
97.00
Add to cart
3-(4-Morpholinyl)propyl isothiocyanate, 96%, Thermo Scientific Chemicals
Catalog numberL12819.06
Price (USD)/ Each
97.00
-
Add to cart
Chemical Identifiers
CAS32813-50-6
IUPAC Name4-(3-isothiocyanatopropyl)morpholine
Molecular FormulaC8H14N2OS
InChI KeyBCEFDMYFAAAFPE-UHFFFAOYSA-N
SMILESS=C=NCCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange to dark brown to brown
Refractive Index1.5280-1.5340 @ 20?C
Assay (GC)≥95.0%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only