Thermo Scientific Chemicals

Divinyl sulfone, 97%, stab. with 0.05% hydroquinone, Thermo Scientific Chemicals

Catalog number: L12827.18
50 g, Each
Thermo Scientific Chemicals

Divinyl sulfone, 97%, stab. with 0.05% hydroquinone, Thermo Scientific Chemicals

Catalog number: L12827.18
50 g, Each
Quantity
Catalog number: L12827.18
also known as L12827-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-77-0
IUPAC Name
(ethenesulfonyl)ethene
Molecular Formula
C4H6O2S
InChI Key
AFOSIXZFDONLBT-UHFFFAOYSA-N
SMILES
C=CS(=O)(=O)C=C
Appearance (Color)
Clear, colorless to yellow to orange
Assay (GC)
≥96.0%
Form
Liquid

Description

Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text